Gene Transcripts

Gene Symbol
Zcchc8
Entrez Gene ID
70650
Gene Name
zinc finger, CCHC domain containing 8
Gene Aliases
5730565F05Rik
Chromosome Location
Chr. 5: 123698302 - 123721044 on Build GRCm38
UniGene ID
Mm.279427
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027494.3 NP_081770.3 10 1108
GenBank mRNA AK017857.1 - 10 1004
AK032349.1 - 10 1085
AK156770.1 - 10 1080
AK166990.1 - 10 1015
AK167126.1 - 10 1013
AK167736.1 - 10 1015
BC013555.1 - 9 760 4238634

View Full Product Details