Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_028343.4 | NP_082619.3 | 9 | 921 | |
GenBank mRNA | AK013269.1 | - | 9 | 895 | |
AK028401.1 | - | 9 | 731 | ||
AK028846.1 | - | 9 | 895 | ||
AK031818.1 | - | 5 | 340 | ||
AK047200.1 | - | 9 | 895 | ||
AK153175.1 | - | 9 | 845 | ||
AY040841.1 | - | 9 | 771 | ||
BC033279.1 | - | 9 | 901 | 4165990 | |
BC050931.1 | - | 9 | 830 | 6811187 |