Gene Transcripts

Gene Symbol
Ndufv2
Entrez Gene ID
72900
Gene Name
NADH dehydrogenase (ubiquinone) flavoprotein 2
Gene Aliases
2900010C23Rik
Chromosome Location
Chr. 17: 66078795 - 66101559 on Build GRCm38
UniGene ID
Mm.2206
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001278415.1 NP_001265344.1 6, 7 650
NM_028388.3 NP_082664.1 5, 6 553
GenBank mRNA AK013511.1 - 5, 6 504
AK078351.1 - 5, 6 497
AK088193.1 - 2, 3 292
AK146998.1 - 5, 6 487
AK150404.1 - 6, 7 587
AK151729.1 - 6, 7 587
AK159460.1 - 5, 6 470
AK166539.1 - 5, 6 514
AK169143.1 - 5, 6 524
AK185171.1 - 1, 2 176
BC030946.1 - 5, 6 482 5003524
CF106105.1 - 4, 5 395

View Full Product Details