Gene Transcripts

Gene Symbol
Dmgdh
Entrez Gene ID
74129
Gene Name
dimethylglycine dehydrogenase precursor
Gene Aliases
1200014D15Rik, AI787269
Chromosome Location
Chr. 13: 93674436 - 93752823 on Build GRCm38
UniGene ID
Mm.21789
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028772.3 NP_083048.1 8 1241
GenBank mRNA AK004755.1 - 8 1242
BC024126.1 - 4 524
BC089599.1 - 8 1244 30118948

View Full Product Details