Gene Transcripts

Gene Symbol
Tm9sf1
Entrez Gene ID
74140
Gene Name
transmembrane 9 superfamily member 1
Gene Aliases
1200014D02Rik, AI893436, MP70
Chromosome Location
Chr. 14: 55635965 - 55643806 on Build GRCm38
UniGene ID
Mm.132857
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028780.3 NP_083056.2 3 811
GenBank mRNA AF269152.1 - 3 459
AK004754.2 - 3 813
AK090065.1 - 2 806
AK160610.1 - 2 803
BC007187.1 - 2 116
BC017617.1 - 2 750 4923337

View Full Product Details