Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_144535.4 | NP_653118.3 | 6 | 1503 | |
GenBank mRNA | AK030928.1 | - | 6 | 1430 | |
AK031527.1 | - | 5 | 1990 | ||
AK041400.1 | - | 6 | 1004 | ||
AK051372.1 | - | 6 | 1450 | ||
AK154166.1 | - | 6 | 1456 | ||
AK164148.1 | - | 6 | 1274 | ||
AK170232.1 | - | 6 | 1280 | ||
AK205351.1 | - | 3 | 164 | ||
AK211028.1 | - | 3 | 156 | ||
BC023094.1 | - | 6 | 1503 | 4017596 |