Gene Transcripts

Gene Symbol
Nipal3
Entrez Gene ID
74552
Gene Name
NIPA-like domain containing 3
Gene Aliases
9130020G22Rik, AI480660, Npal3
Chromosome Location
Chr. 4: 135447136 - 135494806 on Build GRCm38
UniGene ID
Mm.26548
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028995.3 NP_083271.1 11 1279
GenBank mRNA AK018640.1 - 11 1256
AK049696.1 - 11 1314
AK053937.1 - 1283
AK082376.1 - 1281
AK087811.1 - 11 1282
AK138843.1 - 11 1280
AK148206.1 - 11 1301
BC057168.1 - 11 1219 4973200

View Full Product Details