Gene Transcripts

Gene Symbol
4930505A04Rik
Entrez Gene ID
75087
Gene Name
RIKEN cDNA 4930505A04 gene
Gene Aliases
Chromosome Location
Chr. 11: 30426006 - 30471829 on Build GRCm38
UniGene ID
Mm.159422
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100394.1 NP_001093864.1 4 550
GenBank mRNA AK015698.1 - 5 618
BC049719.1 - 4 529

View Full Product Details