Gene Transcripts

Gene Symbol
Lhpp
Entrez Gene ID
76429
Gene Name
phospholysine phosphohistidine inorganic pyrophosphate phosphatase
Gene Aliases
2310007H09Rik
Chromosome Location
Chr. 7: 132610615 - 132706420 on Build GRCm38
UniGene ID
Mm.276721
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029609.1 NP_083885.1 2, 3 386
GenBank mRNA AK009207.1 - 2, 3 384
BC065789.1 - 2, 3 362 832338

View Full Product Details