Gene Transcripts

Gene Symbol
Stambpl1
Entrez Gene ID
76630
Gene Name
STAM binding protein like 1
Gene Aliases
1700095N21Rik, 8230401J17Rik, ALMalpha, AMSH-FP
Chromosome Location
Chr. 19: 34192190 - 34240657 on Build GRCm38
UniGene ID
Mm.130952
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029682.4 NP_083958.3 6 1076
GenBank mRNA AB010121.1 - 6 771
AB010122.1 - 6 735
AB066211.1 - 6 1076
AB066212.1 - 2 425
AB066213.1 - 6 759
AK085665.1 - 5 3467
BC018343.1 - 6 802 4010423
BC023097.1 - 6 1045

View Full Product Details