Gene Transcripts
- Gene Symbol
- Slc10a7
- Entrez Gene ID
- 76775
- Gene Name
- solute carrier family 10 (sodium/bile acid cotransporter family), member 7
- Gene Aliases
- 2410193C02Rik, P7
- Chromosome Location
- Chr. 8: 78509328 - 78734012 on Build GRCm38
- UniGene ID
- Mm.334631
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001009981.2 |
NP_001009981.1
|
7, 8 |
895 |
|
NM_001282109.1 |
NP_001269038.1
|
7, 8 |
903 |
|
NM_029736.2 |
NP_084012.1
|
8, 9 |
979 |
|
GenBank mRNA |
AK010834.1 |
- |
8, 9 |
929 |
|
AK040517.1 |
- |
8, 9 |
858 |
|
AK051650.1 |
- |
5, 6 |
741 |
|
AK152752.1 |
- |
8, 9 |
940 |
|
AK152849.1 |
- |
8, 9 |
940 |
|
AK160402.1 |
- |
8, 9 |
963 |
|
AK165983.1 |
- |
7, 8 |
840 |
|
AY825926.1 |
- |
8, 9 |
732 |
|
AY825927.1 |
- |
7, 8 |
649 |
|
AY825928.1 |
- |
7, 8 |
648 |
|
BC116666.1 |
- |
8, 9 |
845 |
40107077 |
BC116717.1 |
- |
8, 9 |
845 |
40107078 |
View Full Product Details