Gene Transcripts

Gene Symbol
Ralyl
Entrez Gene ID
76897
Gene Name
RALY RNA binding protein-like
Gene Aliases
0710005M24Rik
Chromosome Location
Chr. 3: 13471655 - 14182287 on Build GRCm38
UniGene ID
Mm.422625
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163328.1 NP_001156800.1 5, 6 717
NM_001163329.1 NP_001156801.1 4, 5 310
NM_001163330.1 NP_001156802.1 4, 5 310
NM_178631.4 NP_848746.1 6, 7 810
GenBank mRNA AK032086.1 - 5, 6 650
AK038465.1 - 4, 5 288
AK079659.1 - 5, 6 1539
BC052358.1 - 4, 5 271 6534677
BY252332.1 - 4, 5 311


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA AK003003.1 - 40 Chr. - - [-]

View Full Product Details