Gene Transcripts

Gene Symbol
Armcx1
Entrez Gene ID
78248
Gene Name
armadillo repeat containing, X-linked 1
Gene Aliases
3010033I09Rik, ALEX1
Chromosome Location
Chr. X: 134717912 - 134721917 on Build GRCm38
UniGene ID
Mm.458522
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001166377.1 NP_001159849.1 6 1660
NM_001166378.1 NP_001159850.1 4 1359
NM_001166379.1 NP_001159851.1 4 1475
NM_001166380.1 NP_001159852.1 5 1451
NM_030066.3 NP_084342.1 6 1544
GenBank mRNA AK019405.1 - 6 1471
AK147002.1 - 6 1611
BC021410.1 - 5 1379 5007058
BC026488.1 - 4 1443 4504219
BC068228.1 - 6 1493 6413546

View Full Product Details