Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_019573.3 | NP_062519.2 | 8 | 928 | |
GenBank mRNA | AF187014.1 | - | 8 | 879 | |
AK018507.1 | - | 8 | 858 | ||
AK019911.1 | - | 8 | 886 | ||
AK050541.1 | - | 8 | 880 | ||
AK078528.1 | - | 8 | 881 | ||
BC014716.1 | - | 8 | 884 | 4241066 | |
EF410109.1 | - | 3 | 546 |