Gene Transcripts

Gene Symbol
Chrdl1
Entrez Gene ID
83453
Gene Name
chordin-like 1
Gene Aliases
CHL, CHL1, Nrln1, VOPT
Chromosome Location
Chr. X: 143285674 - 143394262 on Build GRCm38
UniGene ID
Mm.157697
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001114385.1 NP_001107857.1 9 1334
NM_031258.3 NP_112548.2 9 1334
GenBank mRNA AF296451.1 - 9 1256
AF305714.1 - 8 873
AF321853.1 - 8 810
AK142805.1 - 9 958
AK164661.1 - 9 929
BC050818.1 - 8 824
BC066832.1 - 9 968 30102312

View Full Product Details