Gene Transcripts

Gene Symbol
Usp26
Entrez Gene ID
83563
Gene Name
ubiquitin specific peptidase 26
Gene Aliases
Chromosome Location
Chr. X: 51752667 - 51801233 on Build GRCm38
UniGene ID
Mm.242827
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031388.2 NP_113565.2 8 1877
GenBank mRNA AF285570.1 - 5 1473
BC125454.1 - 3 1376 40129969
BC132154.1 - 3 1376 40130431

View Full Product Details