Gene Transcripts

Gene Symbol
Slco1a5
Entrez Gene ID
108096
Gene Name
solute carrier organic anion transporter family, member 1a5
Gene Aliases
Oatp-3, Oatp3, Slc21a7
Chromosome Location
Chr. 6: 142234227 - 142322981 on Build GRCm38
UniGene ID
Mm.206599
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001267707.1 NP_001254636.1 15 2076
NM_130861.3 NP_570931.1 15 2024
GenBank mRNA AF240694.1 - 15 1994
AK029706.1 - 15 2026

View Full Product Details