Gene Transcripts

Gene Symbol
Cnr2
Entrez Gene ID
12802
Gene Name
cannabinoid receptor 2 (macrophage)
Gene Aliases
CB-2, CB2, CB2-R
Chromosome Location
Chr. 4: 135895404 - 135920215 on Build GRCm38
UniGene ID
Mm.297251
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK036658.1 - 2 320
AK037898.1 - 2 430
AK134109.1 - 2 356
BC024052.1 - 2 285 5098695
X86405.1 - 2 273
X93168.1 - 2 341

View Full Product Details