Gene Transcripts

Gene Symbol
Ephb2
Entrez Gene ID
13844
Gene Name
Eph receptor B2
Gene Aliases
Cek5, Drt, ETECK, Erk, Hek5, Nuk, Prkm5, Qek5, Sek3, Tyro5
Chromosome Location
Chr. 4: 136647536 - 136836295 on Build GRCm38
UniGene ID
Mm.250981
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290753.1 NP_001277682.1 15 3006
NM_010142.3 NP_034272.1 15 3003
GenBank mRNA BC043088.1 - 15 2927
BC062924.1 - 15 2951

View Full Product Details