Gene Transcripts

Gene Symbol
Krt13
Entrez Gene ID
16663
Gene Name
keratin 13
Gene Aliases
CK13, K13, Krt-1.13, Krt1-13
Chromosome Location
Chr. 11: 100117596 - 100121495 on Build GRCm38
UniGene ID
Mm.4646
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010662.1 NP_034792.1 3 675
GenBank mRNA AK004635.1 - 3 747
AK009677.1 - 3 236
AK010156.1 - 3 747
AK132640.1 - 3 747
BC060286.1 - 3 690 30292772
X03492.1 - 3 300

View Full Product Details