Gene Transcripts

Gene Symbol
Mknk1
Entrez Gene ID
17346
Gene Name
MAP kinase-interacting serine/threonine kinase 1
Gene Aliases
2410048M24Rik, Mnk1
Chromosome Location
Chr. 4: 115838984 - 115879256 on Build GRCm38
UniGene ID
Mm.209327
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001285487.1 NP_001272416.1 6 563
NM_001285488.1 NP_001272417.1 5 457
NM_021461.5 NP_067436.1 4 413
GenBank mRNA AK010697.1 - 4 391
AK141298.1 - 4 391
AK155934.1 - 6 524
AK169982.1 - 5 423
BC021369.1 - 4 369 5030044
BQ551007.1 - 2 120
Y11091.1 - 4 378

View Full Product Details