Gene Transcripts

Gene Symbol
Slc39a14
Entrez Gene ID
213053
Gene Name
solute carrier family 39 (zinc transporter), member 14
Gene Aliases
FAD-123, ZIP-14, Zip14, fad123
Chromosome Location
Chr. 14: 70303467 - 70351459 on Build GRCm38
UniGene ID
Mm.270647
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135151.1 NP_001128623.1 7 1071
NM_001135152.1 NP_001128624.1 7 1037
NM_144808.4 NP_659057.2 7 1071
GenBank mRNA AB177995.1 - 7 1074
AK090193.1 - 7 1034
AK192726.1 - 2 138
BC021530.1 - 7 1074 5353717
CB521415.1 - 4 399

View Full Product Details