Gene Transcripts

Gene Symbol
Tacr3
Entrez Gene ID
21338
Gene Name
tachykinin receptor 3
Gene Aliases
Nk3r, Nkr, Tac3r
Chromosome Location
Chr. 3: 134829007 - 134934581 on Build GRCm38
UniGene ID
Mm.103810
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021382.6 NP_067357.1 1 532
GenBank mRNA AF233341.1 - 1 266
AK031898.1 - 1 533
BC066845.1 - 1 503 30436441
X87823.1 - 1 490

View Full Product Details