Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_175478.2 | NP_780687.1 | 2 | 1337 | |
GenBank mRNA | AK040916.1 | - | 2 | 1338 | |
BC066999.1 | - | 3 | 1260 | 6418622 | |
BC094593.1 | - | 2 | 1299 | 5708704 | |
DQ078787.1 | - | 1 | 481 |