Gene Transcripts

Gene Symbol
Kcnt2
Entrez Gene ID
240776
Gene Name
potassium channel, subfamily T, member 2
Gene Aliases
E330038N15Rik, Slick
Chromosome Location
Chr. 1: 140245659 - 140613962 on Build GRCm38
UniGene ID
Mm.483845
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081027.2 NP_001074496.2 17 1859
GenBank mRNA AK054538.1 - 5 906

View Full Product Details