Gene Transcripts

Gene Symbol
Olfr1231
Entrez Gene ID
258446
Gene Name
olfactory receptor 1231
Gene Aliases
MOR235-2
Chromosome Location
Chr. 2: 89302649 - 89303590 on Build GRCm38
UniGene ID
Mm.271374
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146454.2 NP_666665.2 1 924
GenBank mRNA BC104282.1 - 1 920
BC104283.1 - 1 923

View Full Product Details