Gene Transcripts
- Gene Symbol
- A130049L09Rik
- Entrez Gene ID
- 320744
- Gene Name
- RIKEN cDNA A130049L09 gene
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK037785.1 |
- |
1 |
1989 |
|
- Gene Symbol
- Zrsr2
- Entrez Gene ID
- 22184
- Gene Name
- zinc finger (CCCH type), RNA binding motif and serine/arginine rich 2
- Gene Aliases
- 35kDa, 5031411E02Rik, A230052C13Rik, C77286, U2af1-rs2, URP
- Chromosome Location
- Chr. X: 163935443 - 163958820 on Build GRCm38
- UniGene ID
- Mm.426178
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK038641.1 |
- |
|
1866 |
|
View Full Product Details