Gene Transcripts

Gene Symbol
Mrgprx1
Entrez Gene ID
404242
Gene Name
MAS-related GPR, member X1
Gene Aliases
MrgC11, Mrgprc11
Chromosome Location
Chr. 7: 48020971 - 48027597 on Build GRCm38
UniGene ID
Mm.354955
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207540.3 NP_997423.1 2 297
GenBank mRNA AY152435.1 - 2 303

View Full Product Details