Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_022419.3 | NP_071864.2 | 2 | 859 | |
GenBank mRNA | AB030191.1 | - | 2 | 849 | |
AK003090.1 | - | 2 | 847 | ||
AK037669.1 | - | 2 | 825 | ||
AK080618.1 | - | 2 | 858 | ||
AK155503.1 | - | 2 | 859 | ||
AK181179.1 | - | 1 | 46 | ||
AK205860.1 | - | 1 | 167 | ||
BC026540.1 | - | 2 | 808 | 5050705 |