Gene Transcripts

Gene Symbol
Cmtm5
Entrez Gene ID
67272
Gene Name
CKLF-like MARVEL transmembrane domain containing 5
Gene Aliases
1500005P16Rik, 2900052H21Rik, Cklfsf5
Chromosome Location
Chr. 14: 54936470 - 54939277 on Build GRCm38
UniGene ID
Mm.41614
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026066.2 NP_080342.1 2 382
GenBank mRNA AK005375.1 - 2 337
AK013666.1 - 2 382
AY241867.1 - 2 372
BC049665.1 - 2 369 6533928

View Full Product Details