Gene Transcripts

Gene Symbol
A1cf
Entrez Gene ID
69865
Gene Name
APOBEC1 complementation factor
Gene Aliases
1810073H04Rik, ACF64, ACF65, ASP, Acf
Chromosome Location
Chr. 19: 31868755 - 31952365 on Build GRCm38
UniGene ID
Mm.87538
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081074.1 NP_001074543.1 10 1535
GenBank mRNA AY566862.1 - 10 1532
AY566863.1 - 10 1508
BC038942.1 - 8 1211

View Full Product Details