Gene Transcripts

Gene Symbol
Ceacam20
Entrez Gene ID
71601
Gene Name
carcinoembryonic antigen-related cell adhesion molecule 20
Gene Aliases
9130012D09Rik
Chromosome Location
Chr. 7: 19965352 - 19991113 on Build GRCm38
UniGene ID
Mm.98097
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027839.2 NP_082115.2 5 968
GenBank mRNA AK018613.1 - 5 965
AK033581.2 - 5 969
BC049361.1 - 5 949 6476373

View Full Product Details