Gene Transcripts
- Gene Symbol
- 1700055D18Rik
- Entrez Gene ID
- 73405
- Gene Name
- RIKEN cDNA 1700055D18 gene
- Gene Aliases
- Chromosome Location
- Chr. 4: 45012559 - 45014356 on Build GRCm38
- UniGene ID
- Mm.82696
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK042394.1 |
- |
1 |
239 |
|
View Full Product Details