Gene Transcripts

Gene Symbol
Thoc3
Entrez Gene ID
73666
Gene Name
THO complex 3
Gene Aliases
2410044K02Rik, AL033344
Chromosome Location
Chr. 13: 54458837 - 54468840 on Build GRCm38
UniGene ID
Mm.292487
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028597.3 NP_082873.2 4 707
GenBank mRNA AK010668.1 - 4 687
BC019603.1 - 4 681 2647864

View Full Product Details