Gene Transcripts

Gene Symbol
Pcdhb2
Entrez Gene ID
93873
Gene Name
protocadherin beta 2
Gene Aliases
PcdhbB
Chromosome Location
Chr. 18: 37294840 - 37297614 on Build GRCm38
UniGene ID
Mm.197508
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053127.2 NP_444357.1 1 1649
GenBank mRNA AY013781.1 - 1 1513
BC021789.1 - 1 1639
BC141154.1 - 1 1605 9055766

View Full Product Details