Gene Transcripts

Gene Symbol
Fam46b
Entrez Gene ID
100342
Gene Name
family with sequence similarity 46, member B
Gene Aliases
4732473B16Rik, AU045678, AU046069
Chromosome Location
Chr. 4: 133480133 - 133487940 on Build GRCm38
UniGene ID
Mm.204656
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175307.6 NP_780516.1 2 737
GenBank mRNA AK028944.1 - 2 734
AK135706.1 - 2 732
AK137659.1 - 2 738
AK162145.1 - 2 445
AK163215.1 - 2 737
BC038001.1 - 2 695 4913445

View Full Product Details