Gene Transcripts

Gene Symbol
Dohh
Entrez Gene ID
102115
Gene Name
deoxyhypusine hydroxylase/monooxygenase
Gene Aliases
1110033C18Rik, AI585913, AL022869, Hlrc1
Chromosome Location
Chr. 10: 81384428 - 81388352 on Build GRCm38
UniGene ID
Mm.294975
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133964.2 NP_598725.2 4 734
GenBank mRNA AK004052.1 - 4 699
AK154104.1 - 4 734
BC002295.1 - 4 651 3590525

View Full Product Details