Gene Transcripts

Gene Symbol
Pycr1
Entrez Gene ID
209027
Gene Name
pyrroline-5-carboxylate reductase 1
Gene Aliases
Chromosome Location
Chr. 11: 120635712 - 120643782 on Build GRCm38
UniGene ID
Mm.127731
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144795.3 NP_659044.1 4 799
GenBank mRNA AK139185.1 - 4 800
BC006727.1 - 5 444 3962093

View Full Product Details