Gene Transcripts

Gene Symbol
Ing5
Entrez Gene ID
66262
Gene Name
inhibitor of growth family, member 5
Gene Aliases
1700001C14Rik, 1700027H23Rik, 1810018M11Rik, AI225768
Chromosome Location
Chr. 1: 93802051 - 93822101 on Build GRCm38
UniGene ID
Mm.64065
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025454.2 NP_079730.1 3 184
GenBank mRNA AK006421.1 - 3 283
AK007536.1 - 3 175
AK009312.1 - 3 168
AK028010.1 - 3 162
AK028397.1 - 3 185
AK145695.1 - 3 157
BC064674.1 - 3 183 6400183

View Full Product Details