Gene Transcripts
- Gene Symbol
- Gm10344
- Entrez Gene ID
- 100038691
- Gene Name
- predicted gene 10344
- Gene Aliases
- ENSMUSG00000071690
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.137751
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK087866.1 |
- |
1 |
525 |
|
View Full Product Details