Gene Transcripts
- Gene Symbol
- Prcp
- Entrez Gene ID
- 72461
- Gene Name
- prolylcarboxypeptidase (angiotensinase C)
- Gene Aliases
- 2510048K03Rik, 2610104A14Rik, AI451719, HUMPCP, PCP
- Chromosome Location
- Chr. 7: 92875253 - 92934581 on Build GRCm38
- UniGene ID
- Mm.389969
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK030160.1 |
- |
|
117 |
|
View Full Product Details