Gene Transcripts

Gene Symbol
Dcun1d4
Entrez Gene ID
100737
Gene Name
DCN1, defective in cullin neddylation 1, domain containing 4 (S. cerevisiae)
Gene Aliases
AI836376
Chromosome Location
Chr. 5: 73481027 - 73560794 on Build GRCm38
UniGene ID
Mm.220312
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001190733.1 NP_001177662.1 7, 8 646
NM_001190734.1 NP_001177663.1 8, 9 581
NM_178896.5 NP_849227.1 7, 8 646
GenBank mRNA AK033559.1 - 7, 8 591
AK076991.1 - 8, 9 582
BC060213.1 - 7, 8 588
BC108346.1 - 7, 8 548 6310392
BC137627.1 - 7, 8 590 8860646
BC137628.1 - 7, 8 590 8860647
BC144794.1 - 7, 8 590 9053342
CO433728.1 - 3, 4 255

View Full Product Details