Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_133927.1 | NP_598688.1 | 4 | 361 | |
GenBank mRNA | AK134630.1 | - | 4 | 382 | |
AK142721.1 | - | 2 | 1892 | ||
AK147728.1 | - | 4 | 363 | ||
AK159637.1 | - | 4 | 370 | ||
AK183090.1 | - | 3 | 106 | ||
AK185688.1 | - | 3 | 105 | ||
BC014833.1 | - | 4 | 361 | 4507661 |