Gene Transcripts

Gene Symbol
Tmem139
Entrez Gene ID
109218
Gene Name
transmembrane protein 139
Gene Aliases
A930027H06Rik
Chromosome Location
Chr. 6: 42261940 - 42264555 on Build GRCm38
UniGene ID
Mm.480678
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175408.4 NP_780617.2 2 281
GenBank mRNA AK053212.1 - 1 1510
AK080737.1 - 1 1328
BY081702.1 - 2 260

View Full Product Details