Gene Transcripts

Gene Symbol
Eya2
Entrez Gene ID
14049
Gene Name
eyes absent 2 homolog (Drosophila)
Gene Aliases
Eab1
Chromosome Location
Chr. 2: 165595028 - 165771727 on Build GRCm38
UniGene ID
Mm.282719
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271962.1 NP_001258891.1 6 507
NM_001271963.1 NP_001258892.1 6 683
NM_010165.3 NP_034295.1 6 684
GenBank mRNA AK137718.1 - 6 679
AK141765.1 - 6 683
AY212512.1 - 5 444
BC003755.1 - 6 684 3497245
CF724561.1 - 5 383
U61111.1 - 5 425
U71208.1 - 6 509
U81603.1 - 5 424

View Full Product Details