Gene Transcripts

Gene Symbol
Gngt1
Entrez Gene ID
14699
Gene Name
guanine nucleotide binding protein (G protein), gamma transducing activity polypeptide 1
Gene Aliases
G(y)1, Gng1
Chromosome Location
Chr. 6: 3989072 - 3997436 on Build GRCm38
UniGene ID
Mm.95398
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010314.2 NP_034444.1 2 94
GenBank mRNA AK020863.1 - 2 96
AK020903.1 - 2 95
BC023064.1 - 2 84
BC025929.1 - 2 289 4511672

View Full Product Details