Gene Transcripts

Gene Symbol
Pax4
Entrez Gene ID
18506
Gene Name
paired box 4
Gene Aliases
Pax-4
Chromosome Location
Chr. 6: 28442334 - 28449340 on Build GRCm38
UniGene ID
Mm.8026
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001159925.1 NP_001153397.1 4 589
NM_001159926.1 NP_001153398.1 4 589
NM_011038.2 NP_035168.1 4 589
GenBank mRNA AB008912.1 - 4 564
AB010557.1 - 4 532
AB010558.1 - 4 547
AF031150.1 - 4 622
AF104231.1 - 3 344
AF145233.1 - 4 536
BC131906.1 - 4 500 40130183
BC131908.1 - 4 498 40130185
BC145168.1 - 4 500 9053719
BC145169.1 - 4 500 9053720
Y09584.1 - 2 228

View Full Product Details