Gene Transcripts

Gene Symbol
Psmb5
Entrez Gene ID
19173
Gene Name
proteasome (prosome, macropain) subunit, beta type 5
Gene Aliases
Chromosome Location
Chr. 14: 54614120 - 54617995 on Build GRCm38
UniGene ID
Mm.8911
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011186.1 NP_035316.1 2 421
GenBank mRNA AF060091.1 - 2 421
AK010441.1 - 2 441
AK133839.1 - 2 442
AK186367.1 - 1 72

View Full Product Details