Gene Transcripts
- Gene Symbol
- Srprb
- Entrez Gene ID
- 20818
- Gene Name
- signal recognition particle receptor, B subunit
- Gene Aliases
- AA409543
- Chromosome Location
- Chr. 9: 103188033 - 103202065 on Build GRCm38
- UniGene ID
- Mm.273053
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_009275.4 |
NP_033301.1
|
5, 6 |
537 |
|
GenBank mRNA |
AK004854.1 |
- |
5, 6 |
570 |
|
AK007859.1 |
- |
5, 6 |
543 |
|
AK008383.1 |
- |
5, 6 |
580 |
|
AK015469.1 |
- |
5, 6 |
602 |
|
AK030437.1 |
- |
5, 6 |
538 |
|
AK075583.1 |
- |
5, 6 |
540 |
|
AK160600.1 |
- |
4, 5 |
1771 |
|
AK167039.1 |
- |
5, 6 |
543 |
|
AK179126.1 |
- |
1, 2 |
90 |
|
AK179649.1 |
- |
1, 2 |
90 |
|
AK181904.1 |
- |
1, 2 |
90 |
|
AK186925.1 |
- |
1, 2 |
90 |
|
AK216962.1 |
- |
1, 2 |
90 |
|
AK219957.1 |
- |
4, 5 |
305 |
|
BC003798.1 |
- |
5, 6 |
558 |
3490766 |
U17343.1 |
- |
5, 6 |
551 |
|
View Full Product Details