Gene Transcripts

Gene Symbol
Aqp12
Entrez Gene ID
208760
Gene Name
aquaporin 12
Gene Aliases
AB084104, mAQP12
Chromosome Location
Chr. 1: 93006334 - 93012269 on Build GRCm38
UniGene ID
Mm.235537
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001159658.1 NP_001153130.1 3 912
NM_177587.2 NP_808255.1 3 915
GenBank mRNA AB084104.1 - 3 978
AK131815.1 - 3 913
BC107212.1 - 3 906 40051607
BC107213.1 - 3 907 40051613

View Full Product Details